Assay Details
| Gene Symbol: LOC101122608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973511.1 | XP_027829312.1 | 12 - 13 | 1119 | 75 | |
| GenBank mRNA | FJ958364.1 | - | 12 - 13 | 918 | 75 | |
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 71 and 85  3' Most  Recommended Assay  Ovis aries | 
 Back To Top
 Back To Top
 LOC101122608
LOC101122608 Genomic Map
Genomic Map Back To Top
 Back To Top