Assay Details
| Gene Symbol: LOC101122608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973511.1 | XP_027829312.1 | 4 - 5 | 451 | 119 | |
| GenBank mRNA | FJ958364.1 | - | 4 - 5 | 250 | 119 | |
More Information
Set Membership: |
|

LOC101122608
Genomic Map