Assay Details
Gene Symbol: LOC101106719 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957647.1 | XP_027813448.1 | 1 - 2 | 383 | 64 | |
XM_027957648.1 | XP_027813449.1 | 2 - 3 | 272 | 64 | ||
XM_027957649.1 | XP_027813450.1 | 1 - 2 | 383 | 64 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
