Assay Details
Gene Symbol: SEC61B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964394.1 | XP_027820195.1 | 3 - 4 | 310 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
