Assay Details
| Gene Symbol: BCL2L11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012173880.3 | XP_012029270.1 | 2 - 3 | 536 | 76 | |
| XM_012173883.3 | XP_012029273.1 | 2 - 3 | 658 | 76 | ||
| XM_027967118.1 | XP_027822919.1 | 2 - 3 | 658 | 76 | ||
| XM_027967119.1 | XP_027822920.1 | 3 - 4 | 490 | 76 | ||
| XM_027967120.1 | XP_027822921.1 | 3 - 4 | 490 | 76 | ||
| GenBank mRNA | HM630307.1 | - | 1 - 2 | 356 | 76 | |
More Information
Set Membership: |
|

BCL2L11
Genomic Map