Assay Details
Gene Symbol: MPHOSPH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004014933.4 | XP_004014982.3 | 4 - 5 | 404 | 86 | |
XM_027977669.1 | XP_027833470.1 | 4 - 5 | 989 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
