Assay Details
| Gene Symbol: GDPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012407.4 | XP_004012456.1 | 4 - 5 | 473 | 102 | |
| XM_027975344.1 | XP_027831145.1 | 4 - 5 | 465 | 102 | ||
More Information
Set Membership: |
|

GDPD1
Genomic Map