Assay Details
| Gene Symbol: DMRT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004342.4 | XP_004004391.2 | 3 - 4 | 809 | 60 | |
| XM_027963916.1 | XP_027819717.1 | 3 - 4 | 815 | 60 | ||
More Information
Set Membership: |
|

DMRT1
Genomic Map