Assay Details
Gene Symbol: PLPPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027969684.1 | XP_027825485.1 | 5 - 6 | 852 | 77 | |
XM_027969686.1 | XP_027825487.1 | 7 - 8 | 1170 | 77 | ||
XM_027969687.1 | XP_027825488.1 | 4 - 5 | 662 | 77 | ||
XM_027969688.1 | XP_027825489.1 | 5 - 6 | 852 | 77 | ||
XM_027969689.1 | XP_027825490.1 | 7 - 8 | 1164 | 77 | ||
XM_027969690.1 | XP_027825491.1 | 7 - 8 | 1247 | 77 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
