Assay Details
| Gene Symbol: TUBB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018984.4 | XP_004019033.1 | 2 - 3 | 369 | 75 | |
| Gene Symbol: LOC443015 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971918.1 | XP_027827719.1 | 1 | 333 | 75 | |
More Information
Set Membership: |
|

TUBB
Genomic Map