Assay Details
| Gene Symbol: TUBB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018984.4 | XP_004019033.1 | 3 - 4 | 475 | 87 | |
| GenBank mRNA | GQ338157.1 | - | 1 - 2 | 112 | 87 | |
| Gene Symbol: LOC443015 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971918.1 | XP_027827719.1 | 1 | 439 | 87 | |
More Information
Set Membership: |
|

TUBB
Genomic Map