Assay Details
| Gene Symbol: GSTM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012178093.3 | XP_012033483.1 | 4 - 5 | 358 | 71 | |
| GenBank mRNA | DQ223552.1 | - | 4 - 5 | 291 | 71 | |
More Information
Set Membership: |
|

GSTM3
Genomic Map