Assay Details
| Gene Symbol: LOC114108999 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027957033.1 | XP_027812834.1 | 1 | 455 | 72 | |
| Gene Symbol: SNX24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015095949.2 | XP_014951435.1 | 6 - 7 | 485 | 72 | |
More Information
Set Membership: |
|

LOC114108999
Genomic Map