Assay Details
| Gene Symbol: PRKAR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142517.1 | NP_001135989.1 | 7 - 8 | 787 | 64 | |
| XM_027973930.1 | XP_027829731.1 | 7 - 8 | 775 | 64 | ||
| XM_027973931.1 | XP_027829732.1 | 7 - 8 | 836 | 64 | ||
| GenBank mRNA | FJ360441.1 | - | 7 - 8 | 787 | 64 | |
More Information
Set Membership: |
|

PRKAR1A
Genomic Map