Assay Details
Gene Symbol: BGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009201.1 | NP_001009201.1 | 3 - 4 | 574 | 66 | |
XM_015104664.2 | XP_014960150.2 | 4 - 5 | 740 | 66 | ||
GenBank mRNA | AF034842.1 | - | 3 - 4 | 574 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
