Assay Details
| Gene Symbol: MYL6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015094795.2 | XP_014950281.1 | 2 - 3 | 262 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYL6B| Gene Symbol: MYL6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015094795.2 | XP_014950281.1 | 2 - 3 | 262 | 75 | |
Set Membership: |
|