Assay Details
Gene Symbol: RNF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003291.4 | XP_004003340.1 | 2 - 3 | 344 | 96 | |
XM_012102263.3 | XP_011957653.1 | 2 - 3 | 284 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
