Assay Details
| Gene Symbol: SMAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017219.4 | XP_004017268.1 | 2 - 3 | 849 | 67 | |
| XM_012097419.3 | XP_011952809.1 | 2 - 3 | 930 | 67 | ||
| XM_015101506.2 | XP_014956992.1 | 2 - 3 | 829 | 67 | ||
| XM_015101507.2 | XP_014956993.1 | 2 - 3 | 920 | 67 | ||
| XM_027956253.1 | XP_027812054.1 | 2 - 3 | 831 | 67 | ||
| GenBank mRNA | AY035385.1 | - | 1 - 2 | 218 | 67 | |
More Information
Set Membership: |
|

SMAD1
Genomic Map