Assay Details
| Gene Symbol: ZDHHC20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015098184.2 | XP_014953670.2 | 4 - 5 | 459 | 79 | |
| XM_027973667.1 | XP_027829468.1 | 4 - 5 | 460 | 79 | ||
| XM_027973668.1 | XP_027829469.1 | 4 - 5 | 459 | 79 | ||
| XM_027973669.1 | XP_027829470.1 | 4 - 5 | 459 | 79 | ||
| XM_027973670.1 | XP_027829471.1 | 4 - 5 | 460 | 79 | ||
| XM_027973671.1 | XP_027829472.1 | 4 - 5 | 460 | 79 | ||
More Information
Set Membership: |
|

ZDHHC20
Genomic Map