Assay Details
| Gene Symbol: LOC114118749 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980149.1 | XP_027835950.1 | 2 - 3 | 591 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC114118749| Gene Symbol: LOC114118749 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980149.1 | XP_027835950.1 | 2 - 3 | 591 | 62 | |
Set Membership: |
|