Assay Details
Gene Symbol: NRBF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004021396.3 | XP_004021445.1 | 3 - 4 | 354 | 66 | |
XM_004021397.3 | XP_004021446.1 | 2 - 3 | 240 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
