Assay Details
Gene Symbol: HCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012189025.2 | XP_012044415.1 | 5 - 6 | 627 | 54 | |
XM_027977061.1 | XP_027832862.1 | 5 - 6 | 670 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() |
