Assay Details
| Gene Symbol: PRRX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015099201.2 | XP_014954687.2 | 2 - 3 | 1511 | 65 | |
| XM_027975594.1 | XP_027831395.1 | 2 - 3 | 1518 | 65 | ||
More Information
Set Membership: |
|

PRRX1
Genomic Map