Assay Details
Gene Symbol: LOC101103154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004009901.4 | XP_004009950.2 | 2 - 3 | 500 | 77 | |
XM_027971062.1 | XP_027826863.1 | 2 - 3 | 492 | 77 | ||
XM_027971063.1 | XP_027826864.1 | 2 - 3 | 492 | 77 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
