Assay Details
Gene Symbol: FBXL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027971208.1 | XP_027827009.1 | 8 - 9 | 1202 | 73 | |
XM_027971209.1 | XP_027827010.1 | 8 - 9 | 1176 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
