Assay Details
Gene Symbol: LOC101110111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027967129.1 | XP_027822930.1 | 6 - 7 | 859 | 75 | |
XM_027967134.1 | XP_027822935.1 | 6 - 7 | 819 | 75 | ||
XM_027967140.1 | XP_027822941.1 | 6 - 7 | 762 | 75 | ||
XM_027967147.1 | XP_027822948.1 | 6 - 7 | 804 | 75 | ||
XM_027967151.1 | XP_027822952.1 | 6 - 7 | 793 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
