Assay Details
Gene Symbol: LOC101121054 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003092.4 | XP_004003141.1 | 15 - 16 | 2125 | 66 | |
XM_027957697.1 | XP_027813498.1 | 14 - 15 | 1721 | 66 | ||
XM_027957708.1 | XP_027813509.1 | 13 - 14 | 1634 | 66 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
