Assay Details
Gene Symbol: LOC101121054 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003092.4 | XP_004003141.1 | 5 - 6 | 948 | 74 | |
XM_027957697.1 | XP_027813498.1 | 4 - 5 | 544 | 74 | ||
XM_027957708.1 | XP_027813509.1 | 3 - 4 | 457 | 74 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
