Assay Details
Gene Symbol: LOC101121054 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003092.4 | XP_004003141.1 | 7 - 8 | 1221 | 99 | |
XM_027957697.1 | XP_027813498.1 | 6 - 7 | 817 | 99 | ||
XM_027957708.1 | XP_027813509.1 | 5 - 6 | 730 | 99 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
