Assay Details
| Gene Symbol: SZRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015093749.2 | XP_014949235.2 | 3 - 4 | 393 | 61 | |
| XM_015093750.2 | XP_014949236.2 | 3 - 4 | 390 | 61 | ||
| XM_027965682.1 | XP_027821483.1 | 2 - 3 | 343 | 61 | ||
| XM_027965683.1 | XP_027821484.1 | 2 - 3 | 340 | 61 | ||
More Information
Set Membership: |
|

SZRD1
Genomic Map