Assay Details
Gene Symbol: LOC101121723 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004019389.4 | XP_004019438.2 | 11 - 12 | 1440 | 84 | |
XM_012101586.3 | XP_011956976.2 | 11 - 12 | 1431 | 84 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
