Assay Details
| Gene Symbol: FITM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014584.4 | XP_004014633.2 | 1 - 2 | 242 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FITM2| Gene Symbol: FITM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014584.4 | XP_004014633.2 | 1 - 2 | 242 | 70 | |
Set Membership: |
|