Assay Details
Gene Symbol: SHARPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011603.3 | XP_004011652.1 | 3 - 4 | 967 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
