Assay Details
Gene Symbol: TINF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012180942.3 | XP_012036332.1 | 1 - 2 | 945 | 55 | |
XM_027971594.1 | XP_027827395.1 | 1 - 2 | 945 | 55 | ||
XM_027971595.1 | XP_027827396.1 | 2 - 3 | 422 | 55 | ||
XM_027971596.1 | XP_027827397.1 | 2 - 3 | 854 | 55 | ||
XM_027971597.1 | XP_027827398.1 | 3 - 4 | 326 | 55 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027971596.1, XM_027971597.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
