Assay Details
| Gene Symbol: C23H18orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960883.1 | XP_027816684.1 | 2 - 3 | 1077 | 67 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C23H18orf25| Gene Symbol: C23H18orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960883.1 | XP_027816684.1 | 2 - 3 | 1077 | 67 | |