Assay Details
| Gene Symbol: MRPS21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002446.4 | XP_004002495.1 | 2 - 3 | 161 | 63 | |
| XM_004002447.3 | XP_004002496.1 | 2 - 3 | 187 | 63 | ||
| XM_004002448.4 | XP_004002497.1 | 2 - 3 | 195 | 63 | ||
| XM_012180483.2 | XP_012035873.1 | 2 - 3 | 206 | 63 | ||
More Information
Set Membership: |
|

MRPS21
Genomic Map