Assay Details
| Gene Symbol: ATP1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009796.1 | NP_001009796.1 | 5 - 6 | 1097 | 65 | |
| XM_027975567.1 | XP_027831368.1 | 4 - 5 | 1054 | 65 | ||
| GenBank mRNA | X03883.1 | - | 1097 | 65 | ||
More Information
Set Membership: |
|

ATP1B1
Genomic Map