Assay Details
Gene Symbol: LOC101106373 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004018986.4 | XP_004019035.1 | 4 - 5 | 1047 | 86 | |
XM_027958869.1 | XP_027814670.1 | 5 - 6 | 1133 | 86 | ||
XM_027958870.1 | XP_027814671.1 | 4 - 5 | 1077 | 86 | ||
XM_027958871.1 | XP_027814672.1 | 4 - 5 | 1008 | 86 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
