Assay Details
Gene Symbol: LOC101106373 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004018986.4 | XP_004019035.1 | 5 - 6 | 1076 | 75 | |
XM_027958869.1 | XP_027814670.1 | 6 - 7 | 1162 | 75 | ||
XM_027958870.1 | XP_027814671.1 | 5 - 6 | 1106 | 75 | ||
XM_027958871.1 | XP_027814672.1 | 5 - 6 | 1037 | 75 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
