Assay Details
Gene Symbol: ALKBH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004023544.4 | XP_004023593.4 | 2 - 3 | 1577 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
