Assay Details
| Gene Symbol: SHOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015092577.2 | XP_014948063.2 | 2 - 3 | 666 | 100 | |
| XM_027959028.1 | XP_027814829.1 | 2 - 3 | 666 | 100 | ||
| XM_027959037.1 | XP_027814838.1 | 3 - 4 | 747 | 100 | ||
More Information
Set Membership: |
|

SHOX2
Genomic Map