Assay Details
| Gene Symbol: BICRAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018824.4 | XP_004018873.2 | 4 - 5 | 4519 | 89 | |
| XM_012100682.2 | XP_011956072.2 | 4 - 5 | 186 | 89 | ||
| XM_015102717.2 | XP_014958203.2 | 4 - 5 | 226 | 89 | ||
| XM_027958602.1 | XP_027814403.1 | 3 - 4 | 5348 | 89 | ||

BICRAL
Genomic Map