Assay Details
| Gene Symbol: MAN1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097327.2 | XP_014952813.2 | 4 - 5 | 955 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAN1A1| Gene Symbol: MAN1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097327.2 | XP_014952813.2 | 4 - 5 | 955 | 69 | |
Set Membership: |
|