Assay Details
Gene Symbol: LOC114115384 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027971374.1 | XP_027827175.1 | 4 | 1115 | 70 |
Gene Symbol: RCCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957077.1 | XP_027812878.1 | 4 - 5 | 1067 | 70 |
