Assay Details
Gene Symbol: LOC101114198 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011124.4 | XP_004011173.2 | 23 - 24 | 3890 | 89 | |
XM_012182449.3 | XP_012037839.2 | 23 - 24 | 3893 | 89 | ||
XM_012182451.3 | XP_012037841.2 | 23 - 24 | 3894 | 89 |
Gene Symbol: LOC114116228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972738.1 | XP_027828539.1 | 1 - 2 | 183 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027972738.1 transcripts.
Set Membership: |
![]() ![]() |
