Assay Details
| Gene Symbol: KIF4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012106845.3 | XP_011962235.1 | 25 - 26 | 2973 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIF4A| Gene Symbol: KIF4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012106845.3 | XP_011962235.1 | 25 - 26 | 2973 | 66 | |
Set Membership: |
|