Assay Details
| Gene Symbol: MAP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015101253.2 | XP_014956739.2 | 5 - 6 | 7298 | 62 | |
| XM_027980016.1 | XP_027835817.1 | 5 - 6 | 7298 | 62 | ||
| XM_027980018.1 | XP_027835819.1 | 4 - 5 | 6986 | 62 | ||
| XM_027980019.1 | XP_027835820.1 | 4 - 5 | 6908 | 62 | ||
More Information
Set Membership: |
|

MAP1B
Genomic Map