Assay Details
| Gene Symbol: GPAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012180288.3 | XP_012035678.1 | 5 - 6 | 888 | 67 | |
| XM_027971150.1 | XP_027826951.1 | 5 - 6 | 963 | 67 | ||
More Information
Set Membership: |
|

GPAT3
Genomic Map