Assay Details
Gene Symbol: LOC114116372 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973017.1 | XP_027828818.1 | 3 - 4 | 558 | 60 | |
XM_027973018.1 | XP_027828819.1 | 3 - 4 | 561 | 60 |
Gene Symbol: LOC101103244 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973014.1 | XP_027828815.1 | 28 - 29 | 3752 | 60 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
