Assay Details
| Gene Symbol: LOC114116372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973017.1 | XP_027828818.1 | 4 - 5 | 645 | 79 | |
| XM_027973018.1 | XP_027828819.1 | 4 - 5 | 648 | 79 | ||
| Gene Symbol: LOC101103244 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973014.1 | XP_027828815.1 | 29 - 30 | 3839 | 79 | |
More Information
Set Membership: |
|

LOC114116372
Genomic Map